Assay Details
| Gene Symbol: LOC102138783 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005540009.2 | XP_005540066.1 | 8 - 9 | 1548 | 105 | |
| XM_015445497.1 | XP_015300983.1 | 8 - 9 | 1548 | 105 | ||
More Information
Set Membership: |
|

LOC102138783
Genomic Map