Assay Details
| Gene Symbol: CHMP2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005590624.2 | XP_005590681.1 | 2 - 3 | 322 | 125 | |
| XM_005590625.2 | XP_005590682.1 | 2 - 3 | 327 | 125 | ||
| XM_015441300.1 | XP_015296786.1 | 2 - 3 | 323 | 125 | ||
| XM_015441301.1 | XP_015296787.1 | 2 - 3 | 328 | 125 | ||
| XM_015441303.1 | XP_015296789.1 | 2 - 3 | 390 | 125 | ||
More Information
Set Membership: |
|

CHMP2A
Genomic Map