Assay Details
Gene Symbol: SULT1B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005555154.2 | XP_005555211.1 | 2 - 3 | 461 | 151 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
