Assay Details
| Gene Symbol: MAD2L1BP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005552900.2 | XP_005552957.1 | 2 - 3 | 396 | 141 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MAD2L1BP| Gene Symbol: MAD2L1BP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005552900.2 | XP_005552957.1 | 2 - 3 | 396 | 141 | |
Set Membership: |
|