Assay Details
| Gene Symbol: MON1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284622.1 | NP_001271551.1 | 5 - 6 | 1517 | 69 | |
| XM_005592605.2 | XP_005592662.1 | 5 - 6 | 1738 | 69 | ||
| XM_005592606.2 | XP_005592663.1 | 5 - 6 | 1728 | 69 | ||
| XM_015442704.1 | XP_015298190.1 | 4 - 5 | 1989 | 69 | ||
| GenBank mRNA | AB178970.1 | - | 5 - 6 | 1517 | 69 | |
More Information
Set Membership: |
|

MON1B
Genomic Map