Assay Details
| Gene Symbol: BAHD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005559182.2 | XP_005559239.2 | 5 - 6 | 2207 | 93 | |
| XM_005559183.2 | XP_005559240.1 | 7 - 8 | 3655 | 93 | ||
| XM_005559184.2 | XP_005559241.1 | 6 - 7 | 3563 | 93 | ||
| XM_015452652.1 | XP_015308138.1 | 5 - 6 | 5163 | 93 | ||
| XM_015452653.1 | XP_015308139.1 | 5 - 6 | 5161 | 93 | ||
| XM_015452655.1 | XP_015308141.1 | 7 - 8 | 2469 | 93 | ||
| XM_015452656.1 | XP_015308142.1 | 5 - 6 | 2310 | 93 | ||
More Information
Set Membership: |
|

BAHD1
Genomic Map