Assay Details
| Gene Symbol: PDZRN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005547625.2 | XP_005547682.1 | 6 - 7 | 1526 | 115 | |
| XM_015446197.1 | XP_015301683.1 | 4 - 5 | 5735 | 115 | ||
| XM_015446198.1 | XP_015301684.1 | 4 - 5 | 1989 | 115 | ||
| XM_015446199.1 | XP_015301685.1 | 4 - 5 | 1594 | 115 | ||
More Information
Set Membership: |
|

PDZRN3
Genomic Map