Assay Details
| Gene Symbol: SYNM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005560577.2 | XP_005560634.1 | 2 - 3 | 1091 | 106 | |
| XM_005560578.2 | XP_005560635.1 | 2 - 3 | 1090 | 106 | ||
More Information
Set Membership: |
|

SYNM
Genomic Map