Assay Details
| Gene Symbol: CNRIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005575707.2 | XP_005575764.1 | 2 - 3 | 1043 | 154 | |
| XM_015433582.1 | XP_015289068.1 | 3 - 4 | 738 | 154 | ||
More Information
Set Membership: |
|

CNRIP1
Genomic Map