Assay Details
| Gene Symbol: ZZZ3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005542971.2 | XP_005543028.1 | 10 - 11 | 2639 | 110 | |
| XM_005542972.2 | XP_005543029.1 | 10 - 11 | 2607 | 110 | ||
| XM_005542974.2 | XP_005543031.1 | 11 - 12 | 2752 | 110 | ||
| XM_005542975.2 | XP_005543032.1 | 11 - 12 | 2671 | 110 | ||
| XM_005542976.2 | XP_005543033.1 | 9 - 10 | 2483 | 110 | ||
| XM_005542977.2 | XP_005543034.1 | 10 - 11 | 2636 | 110 | ||
| XM_015433889.1 | XP_015289375.1 | 9 - 10 | 1083 | 110 | ||
| XM_015433891.1 | XP_015289377.1 | 10 - 11 | 1196 | 110 | ||
| XM_015433894.1 | XP_015289380.1 | 10 - 11 | 1113 | 110 | ||
More Information
Set Membership: |
|

ZZZ3
Genomic Map