Assay Details
| Gene Symbol: ACOT11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005543275.2 | XP_005543332.1 | 13 - 14 | 1545 | 86 | |
| XM_005543276.2 | XP_005543333.1 | 12 - 13 | 1461 | 86 | ||
| XM_005543277.2 | XP_005543334.1 | 13 - 14 | 1667 | 86 | ||
More Information
Set Membership: |
|

ACOT11
Genomic Map