Assay Details
| Gene Symbol: CALY | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005566789.2 | XP_005566846.1 | 3 - 4 | 415 | 121 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CALY| Gene Symbol: CALY | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005566789.2 | XP_005566846.1 | 3 - 4 | 415 | 121 | |
Set Membership: |
|