Assay Details
| Gene Symbol: CHST15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005566677.2 | XP_005566734.1 | 2 - 3 | 1142 | 109 | |
| XM_005566678.2 | XP_005566735.1 | 2 - 3 | 1270 | 109 | ||
| XM_005566679.2 | XP_005566736.1 | 2 - 3 | 1070 | 109 | ||
| XM_015456825.1 | XP_015312311.1 | 1 - 2 | 5289 | 109 | ||
| XM_015456826.1 | XP_015312312.1 | 2 - 3 | 2688 | 109 | ||
More Information
Set Membership: |
|

CHST15
Genomic Map