Assay Details
| Gene Symbol: LOC102123375 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005550913.2 | XP_005550970.1 | 1 - 2 | 226 | 89 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102123375| Gene Symbol: LOC102123375 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005550913.2 | XP_005550970.1 | 1 - 2 | 226 | 89 | |