Assay Details
| Gene Symbol: SCARA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005562918.2 | XP_005562975.1 | 5 - 6 | 1690 | 129 | |
| XM_015454613.1 | XP_015310099.1 | 5 - 6 | 1467 | 129 | ||
More Information
Set Membership: |
|

SCARA3
Genomic Map