Assay Details
| Gene Symbol: TRMO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005581223.2 | XP_005581280.1 | 3 - 4 | 475 | 105 | |
| XM_005581225.2 | XP_005581282.1 | 3 - 4 | 475 | 105 | ||
| XM_015436958.1 | XP_015292444.1 | 3 - 4 | 566 | 105 | ||
| XM_015436959.1 | XP_015292445.1 | 3 - 4 | 663 | 105 | ||
More Information
Set Membership: |
|

TRMO
Genomic Map