Assay Details
| Gene Symbol: ERGIC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285370.1 | NP_001272299.1 | 9 - 10 | 750 | 96 | |
| XM_015430150.1 | XP_015285636.1 | 9 - 10 | 868 | 96 | ||
| GenBank mRNA | AB169621.1 | - | 9 - 10 | 750 | 96 | |
More Information
Set Membership: |
|

ERGIC2
Genomic Map