Assay Details
| Gene Symbol: ECSIT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001319410.1 | NP_001306339.1 | 2 - 3 | 182 | 135 | |
| XM_005588055.2 | XP_005588112.1 | 2 - 3 | 285 | 135 | ||
| XM_015440740.1 | XP_015296226.1 | 3 - 4 | 310 | 135 | ||
| XM_015440741.1 | XP_015296227.1 | 2 - 3 | 347 | 135 | ||
| XM_015440742.1 | XP_015296228.1 | 2 - 3 | 226 | 135 | ||
| GenBank mRNA | AB169489.1 | - | 2 - 3 | 182 | 135 | |
More Information
Set Membership: |
|

ECSIT
Genomic Map