Assay Details
| Gene Symbol: SLC15A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005577696.2 | XP_005577753.1 | 6 - 7 | 2048 | 153 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SLC15A3| Gene Symbol: SLC15A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005577696.2 | XP_005577753.1 | 6 - 7 | 2048 | 153 | |
Set Membership: |
|