Assay Details
| Gene Symbol: CLIC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005552827.1 | XP_005552884.1 | 2 - 3 | 753 | 99 | |
| XM_005552828.2 | XP_005552885.1 | 2 - 3 | 753 | 99 | ||
| XM_005552829.1 | XP_005552886.1 | 2 - 3 | 512 | 99 | ||
| XM_015449346.1 | XP_015304832.1 | 2 - 3 | 1908 | 99 | ||
More Information
Set Membership: |
|

CLIC5
Genomic Map