Assay Details
| Gene Symbol: TRIM44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005578182.2 | XP_005578239.1 | 1 - 2 | 1087 | 81 | |
| XM_005578183.2 | XP_005578240.1 | 1 - 2 | 1087 | 81 | ||
| XM_015434915.1 | XP_015290401.1 | 1 - 2 | 1087 | 81 | ||
More Information
Set Membership: |
|

TRIM44
Genomic Map