Assay Details
| Gene Symbol: MIOX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005566895.2 | XP_005566952.1 | 6 - 7 | 599 | 110 | |
| XM_005566896.2 | XP_005566953.1 | 6 - 7 | 580 | 110 | ||
More Information
Set Membership: |
|

MIOX
Genomic Map