Assay Details
| Gene Symbol: MED29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005589178.2 | XP_005589235.1 | 2 - 3 | 421 | 120 | |
| GenBank mRNA | AB173118.1 | - | 2 - 3 | 287 | 120 | |
More Information
Set Membership: |
|

MED29
Genomic Map