Assay Details
| Gene Symbol: ZNF280C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005594561.2 | XP_005594618.1 | 4 - 5 | 445 | 109 | |
| XM_005594562.2 | XP_005594619.1 | 4 - 5 | 440 | 109 | ||
More Information
Set Membership: |
|

ZNF280C
Genomic Map