Assay Details
| Gene Symbol: SNRK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005546810.1 | XP_005546867.1 | 5 - 6 | 1276 | 112 | |
| XM_005546811.2 | XP_005546868.1 | 6 - 7 | 1326 | 112 | ||
More Information
Set Membership: |
|

SNRK
Genomic Map