Assay Details
| Gene Symbol: PDPR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005592539.2 | XP_005592596.1 | 7 - 8 | 1452 | 111 | |
| XM_015442802.1 | XP_015298288.1 | 6 - 7 | 845 | 111 | ||
| XM_015442803.1 | XP_015298289.1 | 8 - 9 | 1246 | 111 | ||
| XM_015442804.1 | XP_015298290.1 | 6 - 7 | 733 | 111 | ||
More Information
Set Membership: |
|

PDPR
Genomic Map