Assay Details
| Gene Symbol: CDCA8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005588541.1 | XP_005588598.1 | 8 - 9 | 767 | 78 | |
| XM_015440728.1 | XP_015296214.1 | 7 - 8 | 864 | 78 | ||
More Information
Set Membership: |
|

CDCA8
Genomic Map