Assay Details
| Gene Symbol: SEC61A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005564628.2 | XP_005564685.1 | 2 - 3 | 171 | 149 | |
| XM_015455550.1 | XP_015311036.1 | 2 - 3 | 801 | 149 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015455550.1 transcripts.
Set Membership: |
|

SEC61A2
Genomic Map