Assay Details
| Gene Symbol: TMLHE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005595058.2 | XP_005595115.1 | 7 - 8 | 1308 | 80 | |
| XM_005595059.2 | XP_005595116.1 | 8 - 9 | 1437 | 80 | ||
| XM_005595060.2 | XP_005595117.1 | 5 - 6 | 961 | 80 | ||
| XM_005595061.2 | XP_005595118.1 | 6 - 7 | 1125 | 80 | ||
More Information
Set Membership: |
|

TMLHE
Genomic Map