Assay Details
| Gene Symbol: CCSER2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005565267.2 | XP_005565324.1 | 10 - 11 | 2663 | 64 | |
| XM_015456109.1 | XP_015311595.1 | 7 - 8 | 915 | 64 | ||
More Information
Set Membership: |
|

CCSER2
Genomic Map