Assay Details
| Gene Symbol: ELP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005578320.2 | XP_005578377.1 | 1 - 2 | 270 | 92 | |
| XM_005578321.2 | XP_005578378.1 | 1 - 2 | 270 | 92 | ||
| XM_005578322.2 | XP_005578379.1 | 1 - 2 | 269 | 92 | ||
| XM_015434947.1 | XP_015290433.1 | 1 - 2 | 270 | 92 | ||
| XM_015434948.1 | XP_015290434.1 | 1 - 2 | 270 | 92 | ||
| XM_015434949.1 | XP_015290435.1 | 1 - 2 | 250 | 92 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015434949.1 transcripts.
Set Membership: |
|

ELP4
Genomic Map