Assay Details
| Gene Symbol: FAM193B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005558662.2 | XP_005558719.1 | 10 - 11 | 2764 | 103 | |
| XM_005558663.2 | XP_005558720.1 | 9 - 10 | 3451 | 103 | ||
| XM_005558664.2 | XP_005558721.1 | 10 - 11 | 3512 | 103 | ||
| XM_005558666.2 | XP_005558723.1 | 10 - 11 | 3363 | 103 | ||
| XM_015452320.1 | XP_015307806.1 | 10 - 11 | 2659 | 103 | ||
| XM_015452321.1 | XP_015307807.1 | 9 - 10 | 2525 | 103 | ||
| XM_015452323.1 | XP_015307809.1 | 10 - 11 | 3617 | 103 | ||
| XM_015452324.1 | XP_015307810.1 | 9 - 10 | 3556 | 103 | ||
More Information
Set Membership: |
|

FAM193B
Genomic Map