Assay Details
| Gene Symbol: CNNM4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005575003.2 | XP_005575060.1 | 2 - 3 | 1586 | 78 | |
| XM_005575004.2 | XP_005575061.2 | 2 - 3 | 1586 | 78 | ||
More Information
Set Membership: |
|

CNNM4
Genomic Map