Assay Details
| Gene Symbol: FGF22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005587315.2 | XP_005587372.1 | 1 - 2 | 250 | 108 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

FGF22| Gene Symbol: FGF22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005587315.2 | XP_005587372.1 | 1 - 2 | 250 | 108 | |
Set Membership: |
|