Assay Details
| Gene Symbol: RAB22A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005569434.2 | XP_005569491.1 | 4 - 5 | 545 | 84 | |
| XM_015429997.1 | XP_015285483.1 | 3 - 4 | 224 | 84 | ||
More Information
Set Membership: |
|

RAB22A
Genomic Map