Assay Details
| Gene Symbol: SHROOM4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005593588.2 | XP_005593645.1 | 1 - 2 | 150 | 151 | |
| XM_015443473.1 | XP_015298959.1 | 1 - 2 | 552 | 151 | ||
More Information
Set Membership: |
|

SHROOM4
Genomic Map