Assay Details
| Gene Symbol: ZNF608 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005557641.2 | XP_005557698.1 | 9 - 10 | 5000 | 90 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF608| Gene Symbol: ZNF608 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005557641.2 | XP_005557698.1 | 9 - 10 | 5000 | 90 | |
Set Membership: |
|