Assay Details
Gene Symbol: CCDC191 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001284022.1 | NP_001270951.1 | 8 - 9 | 1179 | 93 | |
XM_015445351.1 | XP_015300837.1 | 8 - 9 | 1239 | 93 | ||
GenBank mRNA | AB062941.1 | - | 8 - 9 | 1179 | 93 | |
AB071070.1 | - | 8 - 9 | 1161 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
