Assay Details
| Gene Symbol: FAM234B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005570195.2 | XP_005570252.1 | 12 - 13 | 2073 | 134 | |
| XM_015430396.1 | XP_015285882.1 | 12 - 13 | 2286 | 134 | ||
More Information
Set Membership: |
|

FAM234B
Genomic Map