Assay Details
| Gene Symbol: LOC102122033 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005554583.2 | XP_005554640.1 | 1 - 2 | 160 | 67 | |
| XM_005554584.2 | XP_005554641.1 | 1 - 2 | 160 | 67 | ||
| XM_015450214.1 | XP_015305700.1 | 1 - 2 | 160 | 67 | ||
More Information
Set Membership: |
|

LOC102122033
Genomic Map