Assay Details
| Gene Symbol: MST1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001319531.1 | NP_001306460.1 | 9 - 10 | 1130 | 144 | |
| XM_005547148.2 | XP_005547205.1 | 8 - 9 | 1102 | 144 | ||
| XM_015445912.1 | XP_015301398.1 | 8 - 9 | 1473 | 144 | ||
| GenBank mRNA | AK240624.1 | - | 9 - 10 | 1130 | 144 | |
More Information
Set Membership: |
|

MST1
Genomic Map