Assay Details
| Gene Symbol: NFS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005568875.2 | XP_005568932.1 | 8 - 9 | 1298 | 109 | |
| XM_005568876.2 | XP_005568933.1 | 8 - 9 | 1024 | 109 | ||
| XM_005568878.2 | XP_005568935.2 | 8 - 9 | 1004 | 109 | ||
| XM_015458003.1 | XP_015313489.1 | 8 - 9 | 909 | 109 | ||
More Information
Set Membership: |
|

NFS1
Genomic Map