Assay Details
| Gene Symbol: TRIB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005568595.2 | XP_005568652.1 | 2 - 3 | 925 | 131 | |
| XM_005568596.2 | XP_005568653.1 | 2 - 3 | 569 | 131 | ||
More Information
Set Membership: |
|

TRIB3
Genomic Map