Assay Details
| Gene Symbol: PELI2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005561336.2 | XP_005561393.2 | 2 - 3 | 3318 | 82 | |
| XM_015453815.1 | XP_015309301.1 | 2 - 3 | 5638 | 82 | ||
| XM_015453816.1 | XP_015309302.1 | 2 - 3 | 3334 | 82 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015453816.1 transcripts.
Set Membership: |
|

PELI2
Genomic Map