Assay Details
Gene Symbol: BACH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005552338.2 | XP_005552395.1 | 5 - 6 | 9170 | 143 | |
XM_005552339.2 | XP_005552396.1 | 7 - 8 | 2821 | 143 | ||
XM_005552349.2 | XP_005552406.1 | 6 - 7 | 2742 | 143 | ||
XM_005552350.2 | XP_005552407.1 | 4 - 5 | 9090 | 143 | ||
XM_015449064.1 | XP_015304550.1 | 8 - 9 | 4993 | 143 | ||
XM_015449065.1 | XP_015304551.1 | 8 - 9 | 4998 | 143 | ||
XM_015449066.1 | XP_015304552.1 | 8 - 9 | 2922 | 143 | ||
XM_015449067.1 | XP_015304553.1 | 9 - 10 | 3015 | 143 | ||
XM_015449068.1 | XP_015304554.1 | 8 - 9 | 2917 | 143 | ||
XM_015449069.1 | XP_015304555.1 | 8 - 9 | 2914 | 143 | ||
XM_015449070.1 | XP_015304556.1 | 7 - 8 | 2816 | 143 | ||
XM_015449071.1 | XP_015304557.1 | 5 - 6 | 4003 | 143 | ||
XM_015449073.1 | XP_015304559.1 | 6 - 7 | 2369 | 143 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
