Assay Details
| Gene Symbol: NPAS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005561049.2 | XP_005561106.1 | 3 - 4 | 430 | 115 | |
| XM_015453695.1 | XP_015309181.1 | 3 - 4 | 2197 | 115 | ||
More Information
Set Membership: |
|

NPAS3
Genomic Map