Assay Details
| Gene Symbol: EDAR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005575224.2 | XP_005575281.1 | 7 - 8 | 851 | 93 | |
| XM_005575227.2 | XP_005575284.1 | 7 - 8 | 1093 | 93 | ||
More Information
Set Membership: |
|

EDAR
Genomic Map