Assay Details
| Gene Symbol: HS1BP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005576469.2 | XP_005576526.1 | 4 - 5 | 697 | 57 | |
| XM_005576470.2 | XP_005576527.1 | 4 - 5 | 1046 | 57 | ||
More Information
Set Membership: |
|

HS1BP3
Genomic Map