Assay Details
| Gene Symbol: NMNAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005544840.2 | XP_005544897.1 | 4 - 5 | 1116 | 137 | |
| XM_005544841.2 | XP_005544898.1 | 3 - 4 | 447 | 137 | ||
| XM_015442831.1 | XP_015298317.1 | 4 - 5 | 707 | 137 | ||
More Information
Set Membership: |
|

NMNAT1
Genomic Map