Assay Details
| Gene Symbol: METTL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005587083.2 | XP_005587140.1 | 3 - 4 | 1241 | 129 | |
| GenBank mRNA | AB046105.1 | - | 3 - 4 | 1247 | 129 | |
More Information
Set Membership: |
|

METTL4
Genomic Map